missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ CH1-XL Ligand Leakage ELISA

Product Code. p-7180759 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 Assay
10 Assays
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 18466748

Brand: Thermo Scientific™ 810346201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ CH1-XL Ligand Leakage ELISA Kit is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect CH1-XL Affinity Matrix.

CaptureSelect CH1-XL Ligand Leakage ELISA Kit is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect CH1-XL Affinity Matrix. The ligand, a 14-kD recombinant protein, is covalently coupled for high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect CH1-XL Ligand Leakage ELISA Kit can be used as a tool to aid in development of an optimal purification process and in routine quality control of in-process streams and final product.

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Shipping Condition Dry Ice
Product Line CaptureSelect
Quantity 1 Assay
Type ELISA Kit
Format Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.