missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ CH1-XL RoboColumn™ Pre-packed Column

Product Code. 30276642 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Volume:
200 μL
600 μL
Unit Size:
200µL
600µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Volume Unit Size
30276642 600 μL 600µL
18446578 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30276642 Supplier Thermo Scientific™ Supplier No. 5943462600
  • ONLINE EXCLUSIVE PRICE
    830.00 €
    818.00 € / 600µL
    Save 12.00 €
    1% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ RoboColumn™ small chromatography columns are used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

CaptureSelect RoboColumn™ small chromatography columns are used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

CaptureSelect CH1-XL Affinity Matrix recognizes the CH1 domain of human IgG antibodies, thereby enabling purification of all Fab fragments, independent of the light-chain type (kappa or lambda) and without binding of over-expressed light chain and light-chain dimers. In addition, it can be used for the purification of human IgGs from all four subclasses (1, 2, 3, and 4).

Features of this affinity matrix include:

  • Platform approach for purification of high quality and correctly assembled human Fab fragments
  • No co-purification of over-expressed free light chains due to selectivity for the CH1 domain
  • Elution possible at pH 4.0–4.5 with 50 mM acetate buffer
  • Caustic cleaning with up to 50 mM NaOH
  • Excellent scalability
  • No cross-binding to bovine IgG, therefore presence of fetal bovine serum is not a problem
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Characteristics

  • Column size: 200 μL and 600 μL, 1 strip, 8 columns (8-row)
  • Column dimension: 200 μL: 5 x 10 mm
  • Column dimension: 600 μL: 5 x 30 mm
  • Matrix: agarose based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect CH1-XL affinity ligand
  • Ligand coupling: epoxide coupling
  • Binding capacity: >15 mg/mL human Fab fragment depending on flow rate, column height, contact time. For humanized Fabs, lower capacities may be observed.
  • Elution conditions: 100 mM glycine buffer, pH 3.0; 50 mM acetate buffer, pH 4.0–4.5
  • Formulation buffer: 20% (v/v) ethanol
  • Operating pressure: 2 bar (maximum)
  • Flow rate: 16–150 cm/h

RoboColumn is a trademark of Repligen.

Specifications

Flow Rate 16 to 150 cm/hr.
Max. Pressure 2 bar
Pore Size Not Applicable
Stationary Phase CH1-XL affinity ligand on agarose bead
Column Capacity 600 μL
Length (Metric) 3 cm
Quantity 1 Strip, 8 Columns
Particle Size 65 μm
For Use With (Equipment) Robotic Fluid Handlers
For Use With (Application) Chromatography
Type Affinity Column
Diameter (Metric) Inner 0.5 cm
Content And Storage Store at 2°C to 8°C. Protect from light. Do not freeze.
Binding Capacity >15 mg/mL human Fab fragment
Product Line CaptureSelect
Ligand Type CaptureSelect CH1-XL Affinity Ligand
Matrix Agarose
Shipping Condition Wet Ice
Column Format Pre-packed RoboColumn
Bed Volume 600 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.