missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ GoPure™ FcXP Screening Plate, 96-well, 20 μL

Product Code. 30131704 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Plate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30131704 1 Plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30131704 Supplier Thermo Scientific™ Supplier No. A65813
  • ONLINE EXCLUSIVE PRICE
    1155.00 €
    1110.00 € / Pack of 1
    Save 45.00 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ GoPure™ FcXP 96-well screening plates contain CaptureSelect FcXP Affinity Matrix and provide a versatile platform for sample preparation, handling, and analysis.

CaptureSelect GoPure FcXP 96-well screening plates contain CaptureSelect FcXP Affinity Matrix and provide a versatile platform for sample preparation, handling, and analysis. They accommodate multiple samples simultaneously to achieve high throughput and data generation.

These pre-filled screening plates have the following features:

  • High-throughput screening capability enabling multiple conditions to be evaluated in a single plate experiment
  • 20 μL per well, helping preserve sample while maximizing data generation
  • Ready-to-use 96-well format for efficient evaluation of Thermo Scientific resins

Specifications

Concentration 220 μL Resin Slurry/Well
Content And Storage Store at 2°C to 8°C. Do not freeze.
Shipping Condition Wet Ice
Particle Size 65 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 1 Plate
For Use With (Application) Chromatography
Type Affinity Screening Plate
Sample Volume 20 μL
Ligand Type CaptureSelect FcXP Affinity Ligand
Format 96-well Filter Plate
Capacity (Metric) 800 μL
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.