missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ HRP Anti-AAVX Conjugate

Product Code. 18436658 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
Unit Size:
100µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18436658 100 μL 100µL
30484862 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18436658 Supplier Thermo Scientific™ Supplier No. 7303522100
  • ONLINE EXCLUSIVE PRICE
    3940.00 €
    3785.00 € / 100µL
    Save 155.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ HRP Anti-AAVX Conjugate includes a 14-kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds adeno-associated virus particles of multiple AAV serotypes.

CaptureSelect HRP Anti-AAVX Conjugate includes a 14-kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds adeno-associated virus particles of multiple AAV serotypes. The antibody fragment binds with high affinity to AAV particles and demonstrates good binding reactivity towards a variety of AAV serotypes, including AAV1 to AAV8, and AAVrh10. The antibody fragment is chemically conjugated to horseradish peroxidase (HRP) via a spacer of appropriate length to maintain full binding capacity of the antibody fragment.

CaptureSelect HRP Anti-AAVX Conjugate can be combined with CaptureSelect Biotin Anti-AAVX Conjugate (Cat. No. 7103522100) in enzyme linked immunoassays to quantify the total number of AAV capsids. Sandwich ELISAs can be designed using the biotin anti-AAVX conjugate as a capture reagent immobilized on a streptavidin coated surface. HRP anti-AAVX conjugate is then used for detection of the bound AAV capsids. The unique binding properties of this pair of anti-AAVX conjugates can circumvent the need for incorporation of different AAV serotype-specific antibodies for AAV assay development.

For assay development to measure AAV9 total capsids, we recommend CaptureSelect Biotin Anti-AAV9 Conjugate (Cat. No. 7103332100) and CaptureSelect HRP Anti-AAV9 Conjugate (Cat. No. 7303332100).

Product details

  • Format type: HRP, liquid
  • Buffer: PBS, 50% (v/v) glycerol
  • Storage: in freezer (-5 to -30°C)
  • Concentration: 0.5 mg/mL
  • Volume: 0.1 mL
  • Recommended working dilution for sandwich ELISA: 1: 5000 to 1: 10,000 (the optimal working dilution should be determined empirically for each application)

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/mL
Formulation Liquid, in PBS (7.4), 50% glycerol
Content And Storage Store in freezer (-5°C to -30°C).
Form Liquid
Product Line CaptureSelect
Quantity 100 μL
For Use With (Application) Quantitation of multiple AAV capsids
Type Conjugate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.