missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Human IgG-Fc PK Biotin Conjugate

Product Code. 18426588 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18426588 500 μg 500µg
18466438 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18426588 Supplier Thermo Scientific™ Supplier No. 7103322500
  • ONLINE EXCLUSIVE PRICE
    2005.00 €
    1890.00 € / 500µg
    Save 115.00 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ Human IgG-Fc PK Biotin Conjugate consists of a 13 kDa recombinant single domain antibody fragment that specifically binds to the Fc part of all four human IgG subclasses without cross-binding to mouse, rat, rhesus, and cynomolgus monkey IgG.

CaptureSelect Human IgG-Fc PK Biotin Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the Fc part of all four human IgG subclasses without cross-binding to mouse, rat, rhesus, and cynomolgus monkey IgG. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Human IgG-Fc PK Biotin Conjugate allows you to:

  • Detect, quantitate, and characterize all human IgG antibodies (subclasses 1 to 4), recombinant human IgG antibodies, human IgG-derived Fc-fusion proteins in, for instance, non-human plasma and/or serum samples like mouse, rat , rhesus, and cynomolgus monkey, thereby making it extremely suitable for setting up pharmacokinetics (PK) assays

Applications for CaptureSelect Human IgG-Fc PK Biotin Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Form Liquid
Product Line CaptureSelect
Quantity 500 μg
Type Conjugate
Label or Dye Biotin & Other Haptens
Ligand Type Antibody
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.