Learn More
Thermo Scientific™ CaptureSelect™ Human IgG-Fc PK Biotin Conjugate
Shop All Thermo Scientific ProductsDescription
CaptureSelect Human IgG-Fc PK Biotin Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the Fc part of all four human IgG subclasses without cross-binding to mouse, rat, rhesus, and cynomolgus monkey IgG. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
The CaptureSelect Human IgG-Fc PK Biotin Conjugate allows you to:
- Detect, quantitate, and characterize all human IgG antibodies (subclasses 1 to 4), recombinant human IgG antibodies, human IgG-derived Fc-fusion proteins in, for instance, non-human plasma and/or serum samples like mouse, rat , rhesus, and cynomolgus monkey, thereby making it extremely suitable for setting up pharmacokinetics (PK) assays
Applications for CaptureSelect Human IgG-Fc PK Biotin Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Specifications
Specifications
| Content And Storage | Store in freezer (-5°C to -30°C). |
| Form | Liquid |
| Product Line | CaptureSelect |
| Quantity | 100 μg |
| Type | Conjugate |
| Label or Dye | Biotin & Other Haptens |
| Ligand Type | Antibody |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.