missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ IgG-Fc (Multispecies) Affinity Matrix

Product Code. 18466788 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
Unit Size:
10mL
50mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18466788 5 mL 5mL
18416508 10 mL 10mL
18476718 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18466788 Supplier Thermo Scientific™ Supplier No. 2942852005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ IgG-Fc (Multispecies) Affinity Matrix has been specifically designed to purify IgG through high-affinity binding of the IgG Fc region.

CaptureSelect IgG-Fc (Multispecies) Affinity Matrix has been specifically designed to purify IgG through high-affinity binding of the IgG Fc region. This resin can be used for purification of IgG from a broad range of species. The tested species and their specificity are shown in the figure below.

Features of this affinity matrix include:

  • One-step IgG purification with high purity, from crude material
  • Excellent scalability
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect IgG-Fc (multispecies) affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >25 g IgG/L resin tested with human IgG
  • Elution conditions: 0.1 M glycine, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Binding Capacity >25 g IgG/L resin
Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Matrix Agarose-based, epoxide activated
Particle Size 65 ±10 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 5 mL
Type Affinity Matrix
Ligand Type IgG-Fc (multispecies) affinity ligand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.