missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ KappaXP MiniChrom™ Pre-packed columns

Product Code. 18446668 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Volume:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Volume Unit Size
18446668 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18446668 Supplier Thermo Scientific™ Supplier No. 5943212005
  • ONLINE EXCLUSIVE PRICE
    1590.00 €
    1530.00 € / 5mL
    Save 60.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Thermo Scientific™ CaptureSelect™ KappaXP MiniChrom™ column is designed for bench-scale resin screening, process development, and sample preparation.

CaptureSelect KappaXP MiniChrom™ column is designed for bench-scale resin screening, process development, and sample preparation.

This pre-packed column is ready to connect and has the following features:

  • Volume of 5 mL (0.8 x 10 cm)
  • Direct connection to standard chromatography systems
  • Packed with CaptureSelect KappaXP Affinity Matrix, offering high dynamic binding capacity and high resolution over a wide range of flow rates

The 10-cm bed height of the 5-mL column allows initial process development on a bench scale.

CaptureSelect KappaXP matrix recognizes the kappa light chain of human antibodies, enabling purification of all Kappa containing Ig's, Fab, and Fab2 fragments directly from complex sources and feed streams.

Features of this affinity matrix include:

  • Purification of Kappa monoclonal antibodies, Fab, and Fab2 fragments in a single step with high purity and yield
  • Mild elution for Fab fragments and antibodies
  • Human-specific, no binding to bovine antibodies
  • Excellent scalability
  • Excellent pH stability up to 100 mM NaOH (limited exposure)
  • Non-animal derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect KappaXP affinity ligand
  • Ligand coupling method: epoxide coupling
  • Fab binding capacity: 20–30 g IgG Kappa-Fab/liter resin depending on flow rate, column height, and residence time
  • IgG binding capacity: 35–45 g IgG/liter resin depending on flow rate, column height
  • Elution conditions: 20 mM citric or acetic acid, pH 3–4; 100 mM Tris, 1.5 M MgCl2, pH 6
  • Column dimensions: 5 mL (0.8 x 10 cm)
  • Flow characteristics: 75–150 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Flow Rate 75 to 150 cm/hr. (Up to 2 bar)
Max. Pressure 2 bar
Pore Size Not Applicable
Stationary Phase KappaXP Affinity Matrix with KappaXP Ligand
Length (Metric) 10 cm
Quantity 1 Column
Particle Size 65 μm
For Use With (Equipment) Chromatography Systems
Type Affinity Column
Diameter (Metric) Inner 0.8 cm
Content And Storage Store at 2°C to 8°C. Protect from light. Do not freeze.
Product Line CaptureSelect
Ligand Type CaptureSelect KappaXP Affinity Ligand
Matrix Agarose
Shipping Condition Wet Ice
Column Format Pre-packed
Bed Volume 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.