missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ KappaXP RoboColumn™ Pre-packed columns

Product Code. 30274999 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Volume:
200 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Volume Unit Size
30274999 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30274999 Supplier Thermo Scientific™ Supplier No. 5943212200
  • ONLINE EXCLUSIVE PRICE
    554.00 €
    546.00 € / 200µL
    Save 8.00 €
    1% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

CaptureSelect RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

CaptureSelect KappaXP matrix recognizes the kappa light-chain of human antibodies, enabling purification of all Kappa containing Ig's, Fab, and Fab2 fragments directly from complex sources and feed streams.

Features of this affinity matrix include

  • Purification of Kappa monoclonal antibodies, Fab, and Fab2 fragments in a single step with high purity and yield
  • Mild elution for Fab fragments and antibodies
  • Human-specific, no binding to bovine antibodies
  • Excellent scalability
  • Excellent pH stability up to 100 mM NaOH (limited exposure)
  • Non-animal derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Characteristics

  • Column size: 200 μL, 1 strip, 8 columns (8-row)
  • Column dimension: 5-mm diameter, 10-mm bed height
  • Matrix: agarose based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect KappaXP affinity ligand
  • Ligand coupling method: epoxide coupling
  • Fab binding capacity: 20–30 g IgG Kappa-Fab/liter resin depending on flow rate, column height, and residence time
  • IgG binding capacity: 35–45 g IgG/liter resin depending on flow rate, column height, and residence time
  • Elution conditions: 20 mM citric or acetic acid, pH 3–4; 100 mM Tris, 1.5 M MgCl2, pH6
  • Formulation buffer: 20% (v/v) ethanol
  • Max operating pressure: 2 bar
  • Flow rate: 16–150 cm/hr

RoboColumn is a trademark of Repligen.

Specifications

Max. Pressure 2 bar
Pore Size Not Applicable
Stationary Phase KappaXP Affinity Matrix with KappaXP Ligand
Length (Metric) 1 cm
Quantity 1 Strip, 8 Columns
Particle Size 65 μm
For Use With (Equipment) Chromatography Systems
Type Affinity Column
Diameter (Metric) Inner 0.5 cm
Content And Storage Store at 2°C to 8°C.
Product Line CaptureSelect RoboColumn
Ligand Type CaptureSelect KappaXP Affinity Ligand
Matrix Agarose
Shipping Condition Wet Ice
Column Format Pre-packed RoboColumn
Bed Volume 200 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.