missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ LambdaXP MiniChrom™ Pre-packed Column

Product Code. 30276685 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Volume:
1 mL
5 mL
Unit Size:
1mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Volume Unit Size
30276685 1 mL 1mL
30276436 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30276685 Supplier Thermo Scientific™ Supplier No. 5943752001
  • ONLINE EXCLUSIVE PRICE
    517.00 €
    510.00 € / 1mL
    Save 7.00 €
    1% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ LambdaXP MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.

CaptureSelect LambdaXP MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.

These pre-packed columns are ready to connect and have the following features:

  • Dimensions of 1-mL column: 0.8 x 2 cm
  • Dimensions of 5-mL column: 0.8 x 10 cm
  • Direct connection to standard chromatography systems
  • Packed with CaptureSelect LambdaXP Affinity Matrix, offering high dynamic binding capacity

The 1-mL columns can be used for quick screenings of application feasibility and lab-scale purification on a convenient and easy-to-use pre-packed column. The 10-cm bed height of the 5-mL columns allows initial process development on a bench scale.

CaptureSelect LambdaXP affinity matrix recognizes the constant domain of the human lambda light chain enabling purification of all IgG isotypes including IgG, IgA, IgM, IgD, and IgE directly from complex source materials in a robust and efficient purification process with excellent purity obtained in a single step.

Features of the LambdaXP affinity matrix include:

  • Efficient elution of Fab fragments and immunoglobulins containing a lambda light chain, no cross binding to the Fc-region or kappa light chains
  • One-step purification from crude material with high purity and yield
  • Human specific
  • Non–animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect LambdaXP affinity ligand
  • Ligand coupling: epoxide coupling
  • Binding capacity: >30 g IgG/L resin depending on flow rate, column height, and contact time
  • Elution conditions: 25 mM sodium acetate, pH 3.5
  • Flow characteristics: 50–150 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Flow Rate 50 to 150 cm/hr.
Max. Pressure 2 bar
Pore Size Not Applicable
Stationary Phase Agarose bead
Column Capacity 1 mL
Length (Metric) 2 cm
Quantity 1 Column
Particle Size 65 μm
For Use With (Equipment) Chromatography Systems
Type Affinity Column
Diameter (Metric) Inner 0.8 cm
Content And Storage Store at 2°C to 8°C. Protect from light. Do not freeze.
Binding Capacity >30 g IgG/L
Product Line CaptureSelect
Ligand Type CaptureSelect LambdaXP Affinity Ligand
Matrix Agarose
Shipping Condition Wet Ice
Column Format Pre-packed
Bed Volume 1 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.