Learn More
Thermo Scientific™ CaptureSelect™ LC-kappa (mur) Affinity Matrix
Shop All Thermo Scientific ProductsDescription
CaptureSelect LC-kappa (mur) Affinity Matrix enables purification of murine (mouse, rat, and guinea pig) immunoglobulins and Fab fragments without cross reactivity towards bovine immunoglobulins. The ligand is directed towards a unique domain on the constant part of the kappa light chain of immunoglobulins, which makes CaptureSelect LC-kappa (mur) Affinity Matrix a unique resin in terms of specificity.
This matrix can be used for the purification and isolation of kappa light chain-containing murine immunoglobulins and Fab fragments from complex sources like plasma, serum, and cell culture supernatants in a single step. Due to its non-reactivity with bovine Ig, FCS can be applied in growth media without disturbing purification.
Specifications
Specifications
| Content And Storage | Store at 2°C to 8°C. |
| Particle Size | 35 μm |
| Form | Liquid Suspension |
| Product Line | CaptureSelect |
| Quantity | 5 mL |
| Type | Affinity Matrix |
| Stationary Phase | Anti-LC-kappa (mur) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.