missing translation for 'onlineSavingsMsg'
Learn More

Carbachol, 98.3%

Product Code. 15438859
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438859 1 g 1g
15448859 5 g 5g
15458859 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15438859 Supplier Thermo Scientific Alfa Aesar Supplier No. L06674.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

Carbachol, CAS # 51-83-2, is a parasympathomimetic that acts on acetylcholine receptors. | CAS: 51-83-2 | C6H15ClN2O2 | 182.65 g/mol

Carbachol is a cholinergic agonist and is used for the treatment of glaucoma. It is also used as ophthalmic solution (eye drops) during ophthalmic surgery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Carbachol is an ammonium salt and a choline ester that acts on both muscarinic and nicotinic receptors
  • Carbachol activates the sympathetic nervous system; it leads to intracellular increases in Ca2+ and activates associated signaling pathways
  • Carbachol is not degraded by the cholinesterases

Application

  • Carbachol can inhibit apoptotic cell death, making it useful for the study of cell survival and growth in laboratory settings
  • Carbachol can be used for laboratory research of any biological pathway that depends on acetylcholine, including sleep cycles and motor control
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 51-83-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00012011
InChI Key AIXAANGOTKPUOY-UHFFFAOYSA-N
Synonym carbachol, carbamoylcholine chloride, carbachol chloride, carbacholine, carbocholine, jestryl, doryl, carbamylcholine chloride, miostat, isopto carbachol
PubChem CID 5831
ChEBI CHEBI:3385
IUPAC Name 2-(trimethylazaniumyl)ethyl carbamate chloride
SMILES [Cl-].C[N+](C)(C)CCOC(N)=O

Spezifikation

Melting Point ∽210°C (decomposition)
Quantity 1 g
UN Number UN2811
Beilstein 3917459
Sensitivity Hygroscopic
Merck Index 14,1779
Solubility Information Soluble in water and dimethyl sulfoxide.
Formula Weight 182.65
Percent Purity 98.3%
Chemical Name or Material Carbachol
compliance-icons
Produktidentifikator
  • Carbachol
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 2
Gefahrenhinweis
  • H300-Fatal if swallowed.
Sicherheitshinweis
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Ergänzende Informationen
  • MIXTURE LIST-Contain: Carbachol

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.