missing translation for 'onlineSavingsMsg'
Learn More

Carbamazepine 10,11-Epoxide, TRC

Product Code. 17930385
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17930385 10 mg 10mg
17974075 50 mg 50mg
17965035 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17930385 Supplier Toronto Research Chemicals Supplier No. TRCC17585010MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36507-30-9
Molecular Formula C15 H12 N2 O2
Molecular Weight (g/mol) 252.27
Synonym Carbamazepine 10,11-Epoxide, 1a,10b-Dihydro-6H-dibenz[b,f]oxireno[d]azepine-6-carboxamide, 10,11-Dihydro-5H-dibenz[b,f]azepine-5-carboxamide 10,11-epoxide, 10,11-Epoxycarbamazepine, Carbamazepine 10,11-epoxide, Carbamazepine 10,11-oxide, GP 49-023
IUPAC Name 1a,10b-Dihydro-6H-dibenz[b,f]oxireno[d]azepine-6-carboxamide
SMILES NC(=O)N1c2ccccc2C3OC3c4ccccc14

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H12N2O2/c16-15(18)17-11-7-3-1-5-9(11)13-14(19-13)10-6-2-4-8-12(10)17/h1-8,13-14H,(H2,16,18)
Formula Weight 252.0899
Percent Purity >95%
Chemical Name or Material Carbamazepine-10,11-epoxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.