missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carbamazepine, 98%

Product Code. 10327671 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10327671 5 g Glass bottle 5g
10607754 1 g Glass bottle 1g
10337671 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10327671 Supplier Thermo Scientific Chemicals Supplier No. 344360050
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Carbamazepine (Carbamazepen), CAS # 298-46-4, is an aromatic ring of the dibenzoazepine class that acts as a sodium channel blocker. | CAS: 298-46-4 | C15H12N2O | 236.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Carbamazepine is an aromatic ring of the dibenzoazepine class that acts as a sodium channel blocker
  • It is a tricyclic antidepressant-like drug that reduces post-tetanic potentiation

Applications

  • Carbamazepine can be used as a sodium channel blocker and as a glutamate-transporter agonist to reduce neural activity and produce inhibitory effects in the brain
  • Carbamazepine is useful in the research of pain, epilepsy, and manic depression
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-46-4
Molecular Formula C15H12N2O
Molecular Weight (g/mol) 236.27
MDL Number MFCD00005073
InChI Key FFGPTBGBLSHEPO-UHFFFAOYSA-N
Synonym carbamazepine, tegretol, 5h-dibenzo b,f azepine-5-carboxamide, carbamazepen, carbazepine, finlepsin, carbamezepine, equetro, tegretal, biston
PubChem CID 2554
ChEBI CHEBI:3387
SMILES NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12

Specifications

Melting Point 189°C to 192°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 1783
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,acetone,propylene glycol,dmf,slightly soluble in ethanol,glacial acetic acid
IUPAC Name 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide
Formula Weight 236.27
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Carbamazepine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Carbamazepine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.