missing translation for 'onlineSavingsMsg'
Learn More

Carbamazepine, 98%

Product Code. 15444929
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444929 100 g 100g
15434929 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444929 Supplier Thermo Scientific Alfa Aesar Supplier No. J62590.22
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 298-46-4 | C15H12N2O | 236.27 g/mol

Carbamazepine is used as cytochrome P450 3A4 inducing anti-epileptic drug and ligand for the GABA-A receptor benzodiazepine modulatory site. Carbamazepine is typically used for the treatment of seizure disorders and neuropathic pain. It is associated with other medications and used to treat schizophrenia as well as a second line agent in bipolar disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol, acetone, propylene glycol, chloroform, dimethyl sulfoxide, methanol, dimethyl fluoride,ethylene glycol and monoethyl ether. Slightly soluble in glacial acetic acid. Insoluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-46-4
Molecular Formula C15H12N2O
Molecular Weight (g/mol) 236.27
MDL Number MFCD00005073
InChI Key FFGPTBGBLSHEPO-UHFFFAOYSA-N
Synonym carbamazepine, tegretol, 5h-dibenzo b,f azepine-5-carboxamide, carbamazepen, carbazepine, finlepsin, carbamezepine, equetro, tegretal, biston
PubChem CID 2554
ChEBI CHEBI:3387
IUPAC Name 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide
SMILES NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12

Specifications

Melting Point 189°C to 192°C
Color White
Quantity 100 g
Merck Index 14,1781
Solubility Information Soluble in ethanol,acetone,propylene glycol,chloroform,dimethyl sulfoxide,methanol,dimethyl fluoride,ethylene glycol and monoethyl ether. Slightly soluble in glacial acetic acid. Insoluble in water.
Formula Weight 236.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Carbamazepine
compliance-icons compliance-icons
Product Identifier
  • Carbamazepine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Carbamazepine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.