missing translation for 'onlineSavingsMsg'
Learn More

Carbamazepine, TRC

Product Code. 30137434
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137434 25 g 25g
17963135 1 g 1g
17945105 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137434 Supplier Toronto Research Chemicals Supplier No. TRCC17584025G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-46-4
Molecular Formula C15 H12 N2 O
Molecular Weight (g/mol) 236.27
Synonym 5-Carbamoyl-5H-dibenz[b,f]azepine, Amizepin, Biston, CBZ, Calepsin, Carbamazepen, Carbamazepin, Carbamazepine, Carbatrol, Carbazepine, Carbelan, Epitol, Equetro, Finlepsin, G 32883, Geigy 32883, Karbamazepin, Karbelex, Karberol, NSC 169864, Neurotol, Neurotop, Sirtal, Stazepine, Tegretal, Tegretol, Tegretol XR, Telesmin, Timonil
IUPAC Name benzo[b][1]benzazepine-11-carboxamide
SMILES NC(=O)N1c2ccccc2C=Cc3ccccc13

Specifications

Quantity 25 g
InChI Formula InChI=1S/C15H12N2O/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-10H,(H2,16,18)
Formula Weight 236.095
Percent Purity >95%
Chemical Name or Material Carbamazepine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.