missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Carbamoylcholine chloride

Product Code. 18792734
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18792734 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18792734 Supplier R&D Systems™ Supplier No. 2810/100
  • ONLINE EXCLUSIVE PRICE
    144.26 €
    129.15 € / 100mg
    Save 15.11 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Non-selective cholinergic agonist

Cholinergic receptor agonist that is resistant to the action of cholinesterases. Blocks apoptosis in cerebellar granule neurons.
TRUSTED_SUSTAINABILITY

Specifications

CAS 51-83-2
Quantity 100 mg
Product Line Tocris
Molecular Formula C6H15ClN2O2
Formula Weight 182.65
Purity >98%
Molecular Weight (g/mol) 182.65
Chemical Name or Material Carbamoylcholine chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.