missing translation for 'onlineSavingsMsg'
Learn More

Carbamylcholine chloride, 98+%

Product Code. 10331753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10331753 5 g Glass bottle 5g
11390622 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Código de producto. 10331753 Proveedor Thermo Scientific Acros N.º de proveedor 108240050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 51-83-2 | C6H15ClN2O2 | 182.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 51-83-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00012011
InChI Key AIXAANGOTKPUOY-UHFFFAOYSA-N
Synonym carbachol, carbamoylcholine chloride, carbachol chloride, carbacholine, carbocholine, jestryl, doryl, carbamylcholine chloride, miostat, isopto carbachol
PubChem CID 5831
ChEBI CHEBI:3385
IUPAC Name 2-carbamoyloxyethyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CCOC(N)=O

Especificaciones

Melting Point 200°C to 204°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99%
Linear Formula H2NCO2CH2CH2N(CH3)3Cl
Merck Index 15,1781
Solubility Information Solubility in water: 1g/mL. Other solubilities: 1g/50 mL alcohol,1g/10 mL methanol,practically insoluble in chloroform and ether
Formula Weight 182.65
Percent Purity 99%
Infrared Spectrum Authentic
Loss on Drying 2.0% max. (105°C, 2 hrs)
Packaging Glass bottle
Chemical Name or Material Carbamylcholine chloride
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Carbamylcholine chloride
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 2
Indicaciones de peligro
  • H300-Fatal if swallowed.
Consejos de prudencia
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.