missing translation for 'onlineSavingsMsg'
Learn More

Carbamylcholine Chloride, TRC

Product Code. 30362228
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30362228 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30362228

Brand: Toronto Research Chemicals TRCC1759705G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbachol
CAS 51-83-2
Molecular Formula C6 H15 N2 O2 . Cl
Quantity 5 g
Synonym Carbachol, 2-(Carbamoyloxy)-N,N,N-trimethylethanaminium chloride, Choline, chloride, carbamate (6CI,8CI), Ethanaminium, 2-[(aminocarbonyl)oxy]-N,N,N-trimethyl-, chloride (9CI), (2-Carbamoyloxyethyl)trimethylammonium chloride, (2-Hydroxyethyl)trimethyl ammonium chloride carbamate, Carbachol chloride, Carbacholin, Carbacholine, Carbacholine chloride, Carbacolina, Carbaminocholine chloride, Carbaminoylcholine chloride, Carbamiotin, Carbamoylcholine chloride, Carbamoylcholine-hydrochloride, Carbamylcholine chloride, Carbastat Intraocular, Carbochol, Carbocholin, Carbocholine, Carbyl, Carcholin, Choline carbamate chloride, Coletyl, Doryl, Doryl (pharmaceutical), Isopto Carbachol, Jestryl, Lentin, Miostat, Moryl, Vasoperif, Ethanaminium, 2-[(aminocarbonyl)oxy]-N,N,N-trimethyl-, chloride (1:1), gamma-Carbamoyl choline chloride
InChI Formula InChI=1S/C6H14N2O2.ClH/c1-8(2,3)4-5-10-6(7)9;/h4-5H2,1-3H3,(H-,7,9);1H
SMILES [Cl-].C[N+](C)(C)CCOC(=O)N
IUPAC Name 2-carbamoyloxyethyl(trimethyl)azanium;chloride
Molecular Weight (g/mol) 182.65
Formula Weight 182.0822
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.