missing translation for 'onlineSavingsMsg'
Learn More

Carbazole, 96%

Product Code. 11996591 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
1kg
250g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11996591 1 kg Plastic bottle 1kg
10469780 5 g Glass Bottle 5g
10050620 25 g Plastic bottle 25g
10479780 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11996591 Supplier Thermo Scientific Acros Supplier No. 108260010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86-74-8 | C12H9N | 167.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-74-8
Molecular Formula C12H9N
Molecular Weight (g/mol) 167.21
MDL Number MFCD00004960
InChI Key UJOBWOGCFQCDNV-UHFFFAOYSA-N
Synonym carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhn
PubChem CID 6854
ChEBI CHEBI:27543
IUPAC Name 9H-carbazole
SMILES N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 240.0°C to 246.0°C
Color Beige-Brown or Beige-Yellow
Boiling Point 355.0°C
Flash Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Plastic bottle
Quantity 1 kg
Beilstein 20,433
Merck Index 15,1791
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in acetic acid- soluble in h2so4,1g/3mL quinoline- 1g/6mL pyridine- 1g/35mL ether,1g/9mL acetone- 1g/120mL benzene,1g/135mL abs. alcohol- slightly soluble in,petroleum ether and
Formula Weight 167.21
Percent Purity 96%
Physical Form Crystalline Powder, Flakes, or Chunks
Chemical Name or Material Carbazole, 96%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Carbazole
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.