missing translation for 'onlineSavingsMsg'
Learn More

Carbocisteine, TRC

Product Code. 30138751
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138751 5 g 5g
30134998 25 g 25g
17997864 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138751 Supplier Toronto Research Chemicals Supplier No. TRCC1787605G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-23-3
Molecular Formula C5 H9 N O4 S
Molecular Weight (g/mol) 179.19
Synonym Carbocisteine, Alanine, 3-[(carboxymethyl)thio]-, L- (6CI,8CI), (L)-2-Amino-3-(carboxymethylthio)propionic acid, (R)-S-(Carboxymethyl)cysteine, 3-[(Carboxymethyl)thio]-L-alanine, AHR 3053, Bronchokod, Carbocit, Carbocysteine, Chilvax, DF 1794Y, L-(Carboxymethyl)cysteine, LJ 206, Lisil, Lisomucil, Loviscol, Muciclar, Mucocis, Mucodyne, Mucofan, Mucolase, Mucolex, Mucopront, Mucotab, Mukinyl, NSC 14156, Pectox, Pulmoclase, Reomucil, Rhinathiol, Rhinatiol, Rinatiol, S-(Carboxymethyl)-(R)-cysteine, S-(Carboxymethyl)-L-cysteine, S-Carboxylmethyl-L-cysteine, Siroxyl, Thiodril, Transbronchin
IUPAC Name (2R)-2-amino-3-(carboxymethylsulfanyl)propanoic acid
SMILES N[C@@H](CSCC(=O)O)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C5H9NO4S/c6-3(5(9)10)1-11-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1
Formula Weight 179.0252
Percent Purity >95%
Chemical Name or Material Carbocisteine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.