missing translation for 'onlineSavingsMsg'
Learn More

Carbon, activated, 2mm & down

Product Code. 11371458
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11371458 100 g 100g
11351458 250 g 250g
11361458 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11371458 Supplier Thermo Scientific Alfa Aesar Supplier No. 010926.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For liquid purification | CAS: 1333-86-4 | C | 12.01 g/mol

For liquid purification

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1333-86-4
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N

Specifications

Quantity 100 g
Chemical Name or Material Carbon, Activated

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.