missing translation for 'onlineSavingsMsg'
Learn More

Carbon (Amorphous) (13C, 99 atom %), TRC

Product Code. 17924655
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17924655 10 mg 10mg
17941815 100 mg 100mg
17972135 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17924655 Supplier Toronto Research Chemicals Supplier No. TRCC78016010MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14762-74-4
Molecular Formula 13C
Molecular Weight (g/mol) 13
IUPAC Name carbon-13
SMILES [13C]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C/i1+1
Formula Weight 13.0034
Chemical Name or Material Carbon-13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.