missing translation for 'onlineSavingsMsg'
Learn More

Carbon black, acetylene, 50% compressed, 99.9+%

Product Code. 11344687
Change view
Click to view available options
Quantity:
250 g
1 kg
3 kg
Unit Size:
1kg
250g
3kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344687 3 kg 3kg
11354687 250 g 250g
11334687 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11344687 Supplier Thermo Scientific Alfa Aesar Supplier No. 039724.A5
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form:S.A. 75m2/g, Bulk density 80-120g/L | CAS: 1333-86-4 | C | 12.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1333-86-4
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N

Specifications

Color Black
Assay Percent Range >99.9%
Odor Odorless
Quantity 3 kg
Physical Form Powder
Chemical Name or Material Carbon black
What do the "50% compressed" and "100% compressed" designations mean in the description for Carbon black, acetylene?

The designation of compression is a generic description of density via mechanical compression of Carbon black when manufactured. Carbon black, acetylene is difficult to densify due to its high structure and the production process. As a result, compression is used to press air out of the powder and allow the density to be recorded. Based on the percentage of compression (50% or 100%), the density can be recorded and used to generate a higher-than-normal density:

  • Carbon black, acetylene, 100% compressed (Cat. No. 045527.xx) has a density of 135-160 g/L
  • Carbon black, acetylene, 50% compressed, 99.9+% (Cat. No. 039724.xx) has a density of 90-121 g/L

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.