missing translation for 'onlineSavingsMsg'
Learn More

Carbon disulfide, 99.9%, for spectroscopy

Product Code. 10399083 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass Bottle
Unit Size:
2.5 L
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10399083 2.5 L Glass Bottle 2.5 L
10245980 25 mL Glass Bottle 25mL
10561231 500 mL Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10399083 Supplier Thermo Scientific Acros Supplier No. 167710025
  • ONLINE EXCLUSIVE PRICE
    201.00 €
    195.00 € / 2.5 L
    Save 6.00 €
    2% Off
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-15-0 | CS2 | 76.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
MDL Number MFCD00011321
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Melting Point -111.0°C
Absorbance 0.01 max. at 420nm, 0.05 max. at 410nm, 0.1 max. at 400nm, 0.5 max. at 390nm, 1.3 max. at 380nm
Density 1.2620g/mL
Quantity 2.5 L
Flash Point −30°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.627
Residue after Evaporation 0.0005% max.
Assay Percent Range 99.9%
Specific Gravity 1.262
Linear Formula CS2
Viscosity 0.363 mPa.s (20°C)
Water 0.03% max.
Beilstein 03, 197
Fieser 01,114; 05,94; 06,95
Merck Index 15, 1811
Boiling Point 46.0°C
Solubility Information Solubility in water: 2.9g/L (20°C). Other solubilities: miscible with anhydrous methanol, ethanol, ether,, benzene, chloroform, carbon tetrachloride and oils
Formula Weight 76.13
Percent Purity 99.9%
Grade Spectroscopy
Chemical Name or Material Carbon disulfide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Carbon disulfide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P362-Take off contaminated clothing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.