missing translation for 'onlineSavingsMsg'
Learn More

Carbon Disulfide, 99+%, Extra Pure, SLR, Fisher Chemical

Product Code. 11448103 Shop All Fisher Chemical Products
Change view
Click to view available options
2.5L, Plastic Coated Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11448103 2.5L, Plastic Coated Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11448103 Supplier Fisher Chemical Supplier No. C/2880/PB17
Quantity Price
16 81.40 € / 2.5 L
1 95.76 € / 2.5 L
4 86.18 € / 2.5 L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

pH 5
Melting Point -111°C
CAS 75-15-0
Color Colorless
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Molecular Formula CS2
Vapor Pressure 400 hPa at 20°C
MDL Number 11321
Viscosity 0.363cp at 20°C
Boiling Point 46°C
Formula Weight 76.13g/mol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Carbon disufide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 1
  • Reproductive toxicity Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.