missing translation for 'onlineSavingsMsg'
Learn More

Carbon Disulfide, Solstice

Product Code. 15683470
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15683470 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15683470 Supplier Fluka Supplier No. 316271L
  • ONLINE EXCLUSIVE PRICE
    132.00 €
    116.35 € / 1 L
    Save 15.65 €
    11% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for IR spectroscopy, Puriss. p.a., ACS reagent, Reag. Ph. Eur.,≥99.9% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-15-0
Density 1.266g/mL at 25°C
Quantity 1 L
Explosion Limit 0.5
Flash Point −30°C (−22°F)
Ignition Point 212°F
Molecular Formula CS2
MDL Number MFCD00011321
UN Number 1131
Beilstein 1098293
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILES C(=S)=S
IUPAC Name methanedithione
Molecular Weight (g/mol) 76.131
PubChem CID 6348
ChEBI CHEBI:23012
Formula Weight 76.14g/mol
Percent Purity ≥99.9% (GC)
Grade puriss. p.a.
Chemical Name or Material Carbon Disulfide, Reag. Ph. Eur.
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Carbon disulfide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: carbon disulphide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.