Learn More
Swann-Morton™ Carbon Steel Scalpel Blades
Product Code. 11841733
Change view
Click to view available options
Size:
# 14
# 20
# 22A
# 27
# 36
60 x 10.96 mm
Major-Z
Unit Size:
Pack of 100
Pack of 50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11841733 # 36 Pack of 100
11320915 60 x 10.96 mm Pack of 50
11768363 # 20 Pack of 100
12474070 # 27 Pack of 100
11837383 # 14 Pack of 100
12308019 # 22A Pack of 100
12922545 Major-Z Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11841733 Supplier Swann-Morton™ Supplier No. 0236
  • ONLINE EXCLUSIVE PRICE
    31.90 €
    29.01 € / Pack of 100
    Save 2.89 €
    9% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use Swann Morton™ Carbon Steel Scalpel Blades in the operating room for precise cutting and repair.
  • Carbon steel to BS EN 27740
  • Handles and blades sold separately
Specifications
Blade Material Carbon Steel
Product Type Scalpel Blade
For Use With (Application) General Surgery, Laboratory Setting for Histology and Histopathology
Size # 36
Sterility Sterile
Quantity 10/Pk.