missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carbonate, 0.5M buffer soln., pH 9.5

Product Code. 15495829
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Product Code. Quantity unitSize
15495829 500 mL 500mL
15485829 250 mL 250mL
2 options
This item is not returnable. View return policy

Product Code. 15495829

Brand: Thermo Scientific Chemicals J63658.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium bicarbonate buffer system is important for several different cellular processes. It is one of the major buffering systems used to maintain the pH of mammalian blood and is also important in the acid-base homeostasis of living things, including humans. It tends to maintain a relatively constant plasma pH and counteract any force that would alter it. It is used to neutralize the pH of the chyme leaving the stomach and entering the small intestine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bicarbonate buffer system is important for several different cellular processes. It is one of the major buffering systems used to maintain the pH of mammalian blood and is also important in the acid-base homeostasis of living things, including humans. It tends to maintain a relatively constant plasma pH and counteract any force that would alter it. It is used to neutralize the pH of the chyme leaving the stomach and entering the small intestine.

Solubility
Soluble in water.

Notes
Store at 4°C in tightly closed container. Avoid inhalation. Wash thoroughly after handling. Avoid prolonged or repeated exposure.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 M buffer soln.
pH Range 9.5
Quantity 500 mL
Type Carbonate buffer soln.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.