missing translation for 'onlineSavingsMsg'
Learn More

Carboplatin

Product Code. 15493199
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493199 5 g 5g
15483199 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493199 Supplier Thermo Scientific Alfa Aesar Supplier No. J60433.06
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 41575-94-4 | C6H12N2O4Pt | 371.26 g/mol

Carboplatin is primarily used as an antitumor agent.Carboplatin is widely utilized as a chemotherapy drug to treat various types of cancer. It belongs to the group of platinum-based antineoplastic agents to interfere in DNA (deoxyribonucleic acid) repair. It is applied in combination other medications and used to inhibit cancer cell growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41575-94-4
Molecular Formula C6H12N2O4Pt
Molecular Weight (g/mol) 371.26
MDL Number MFCD00070464
InChI Key BHKICZDKIIDMNR-UHFFFAOYSA-L
Synonym carboplatin
PubChem CID 122130863
IUPAC Name azanide;cyclobutane-1,1-dicarboxylate;platinum(4+)
SMILES N.N.O=C1O[Pt++]OC(=O)C11CCC1

Specifications

Color White
Linear Formula [C4H6(CO2)2]Pt(NH3)2
Quantity 5 g
Merck Index 14,1822
Solubility Information Soluble in water.
Formula Weight 371.25
Physical Form Powder
Chemical Name or Material Carboplatin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.