missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Carboplatin
GREENER_CHOICE

Product Code. 18772504
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18772504 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18772504 Supplier Tocris Bioscience™ Supplier No. 2626/50
  • ONLINE EXCLUSIVE PRICE
    186.00 €
    175.35 € / 50mg
    Save 10.65 €
    5% Off
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

DNA cross-linking antitumor agent

Antitumor agent that forms platinum-DNA adducts. Causes intra- and interstrand DNA crosslinks blocking DNA replication and transcription. Enhances radiation-induced single-strand DNA breakage and displays lower nephrotoxicity than analog cisplatin (Cat. No. 2251).
TRUSTED_SUSTAINABILITY

Specifications

CAS 41575-94-4
Quantity 50 mg
Molecular Formula C6H12N2O4Pt
Formula Weight 371.25
Purity >99%
Molecular Weight (g/mol) 371.26
Chemical Name or Material Carboplatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.