missing translation for 'onlineSavingsMsg'
Learn More

Carboprost Tromethamine, TRC

Product Code. 30132463
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132463 25 mg 25mg
30004732 5 mg 5mg
30007020 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132463 Supplier Toronto Research Chemicals Supplier No. TRCC17760025MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58551-69-2
Molecular Formula C21 H36 O5 . C4 H11 N O3
Molecular Weight (g/mol) 489.64
Synonym Carboprost trometamol, 2-Amino-2-(hydroxymethyl)propane-1,3-diol (5Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(1E,3S)-3-hydroxy-3-methyloct-1-enyl]cyclopentyl]hept-5-enoate, (15S)-15-methyl-PGF2
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;(Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(E,3S)-3-hydroxy-3-methyloct-1-enyl]cyclopentyl]hept-5-enoic acid
SMILES CCCCC[C@](C)(O)\C=C\[C@H]1[C@H](O)C[C@H](O)[C@@H]1C\C=C/CCCC(=O)O.NC(CO)(CO)CO

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H36O5.C4H11NO3/c1-3-4-9-13-21(2,26)14-12-17-16(18(22)15-19(17)23)10-7-5-6-8-11-20(24)25;5-4(1-6,2-7)3-8/h5,7,12,14,16-19,22-23,26H,3-4,6,8-11,13,15H2,1-2H3,(H,24,25);6-8H,1-3,5H2/b7-5-,14-12+;/t16-,17-,18+,19-,21+;/m1./s1
Formula Weight 489.3302
Chemical Name or Material Carboprost trometamol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.