missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carborundum, Technical, SLR, Extra Fine Powder, about 36μm (300 Grit), Fisher Chemical
Shop All Fisher Chemical ProductsSpecifications
Specifications
| Physical Form | Solid |
| Packaging | Plastic Powder Jar |
| Color | Black |
| Quantity | 500 g |
| CAS | 409-21-2 |
| Melting Point | 2700°C |
| MDL Number | 49531 |
| Synonym | Silicon carbide |
| Formula Weight | 40.0855g/mol |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?