missing translation for 'onlineSavingsMsg'
Learn More

Carborundum, Technical, SLR, Powder, Coarse, about 686μm (24 Grit), Fisher Chemical

Product Code. 10600341 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10600341 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10600341 Supplier Fisher Chemical Supplier No. C/3090/53
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Specifications

Physical Form Solid
Packaging Plastic Powder Jar
Color Black
Quantity 500 g
CAS 409-21-2
Melting Point 2700°C
MDL Number 49531
Synonym Silicon carbide
Formula Weight 40.0855g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.