missing translation for 'onlineSavingsMsg'
Learn More

(-)-Carbovir, TRC

Product Code. 17992735
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992735 250 mg 250mg
17961315 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17992735 Supplier Toronto Research Chemicals Supplier No. TRCC177685250MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120443-30-3
Molecular Formula C11 H13 N5 O2
Synonym 6H-Purin-6-one, 2-amino-1,9-dihydro-9-[(1R,4S)-4-(hydroxymethyl)-2-cyclopenten-1-yl]-∣6H-Purin-6-one, 2-amino-1,9-dihydro-9-[4-(hydroxymethyl)-2-cyclopenten-1-yl]-, (1R-cis)-∣2-Amino-1,9-dihydro-9-[(1R,4S)-4-(hydroxymethyl)-2-cyclopenten-1-yl]-6H-purin-6-one∣(-)-Carbovir

Specifications

Format Neat
Molecular Weight (g/mol) 247.2532
Product Type API
Shipping Condition Room Temperature
Quantity 250 mg
Chemical Name or Material (-)-Carbovir
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.