missing translation for 'onlineSavingsMsg'
Learn More

Carboxybutyrolactone, TRC

Product Code. 30352632
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30352632 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352632 Supplier Toronto Research Chemicals Supplier No. TRCC1782001G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4344-84-7
Molecular Formula C5H6O4
Quantity 1 g
Synonym Tetrahydro-5-oxo-2-furancarboxylic Acid, 2-Hydroxyglutaric Acid gamma-Lactone, 5-Oxo-2-tetrahydrofurancarboxylic Acid, Butyrolactone-5-carboxylic Acid, Butyrolactonecarboxylic Acid, NSC 72000, 2-Hydroxypentanedioic Acid gamma-Lactone, Tetrahydro-5-oxo-2-furoic Acid, alpha-Hydroxyglutaryl Lactone, gamma-Butyrolactone-gamma-carboxylic Acid
InChI Formula InChI=1S/C5H6O4/c6-4-2-1-3(9-4)5(7)8/h3H,1-2H2,(H,7,8)
SMILES OC(=O)C1CCC(=O)O1
IUPAC Name 5-oxooxolane-2-carboxylic acid
Molecular Weight (g/mol) 130.1
Formula Weight 130.0266
Chemical Name or Material Carboxybutyrolactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.