missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethoxyamine Hemihydrochloride, TRC

Product Code. 30361183
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361183 10 g 10g
30363669 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30361183 Supplier Toronto Research Chemicals Supplier No. TRCC17873010G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2921-14-4
Molecular Formula 2 C2 H5 N O3 . Cl H
Molecular Weight (g/mol) 218.59
Synonym Aminooxyacetic Acid Hemihydrochloride, 2-(Aminooxy)acetic Acid Hemihydrochloride, Carboxymethoxyamine Hemihydrochloride, Carboxymethoxylamine Hemihydrochloride, O-Carboxymethylhydroxylamine Hemihydrochloride
IUPAC Name 2-aminooxyacetic acid;hydrochloride
SMILES Cl.NOCC(=O)O.NOCC(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/2C2H5NO3.ClH/c2*3-6-1-2(4)5;/h2*1,3H2,(H,4,5);1H
Formula Weight 218.0306
Chemical Name or Material Carboxymethoxyamine Hemihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.