missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethoxylamine HemiHydroChloride, 98%

Product Code. 10447290 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10447290 2.5 g Glass bottle 2.5g
10771994 10 g Glass bottle 10g
10165593 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10447290 Supplier Thermo Scientific Acros Supplier No. 108430025
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2921-14-4 | C4H11ClN2O6 | 218.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2921-14-4
Molecular Formula C4H11ClN2O6
Molecular Weight (g/mol) 218.59
MDL Number MFCD00012955
InChI Key AXIKLJPWEGNZIG-UHFFFAOYSA-N
Synonym o-carboxymethyl hydroxylamine hemihydrochloride, carboxymethoxylamine hemihydrochloride, aminooxyacetic acid hemihydrochloride, 2-aminooxy acetic acid hydrochloride 2:1, unii-5g5qi2gn7a, acetic acid, aminooxy-, hydrochloride 2:1, aminooxy acetic acid hemihydrochloride, 5g5qi2gn7a, carboxymethoxyamine hemihydrochloride
PubChem CID 2723609
IUPAC Name 2-aminooxyacetic acid;hydrochloride
SMILES [Cl-].NOCC(O)=O.[NH3+]OCC(O)=O

Specifications

Melting Point 156.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (H2NOCH2CO2H)2·HCl
Quantity 2.5 g
Solubility Information Solubility in water: decomposes.
Formula Weight 109.3
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Carboxymethoxylamine hemihydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.