missing translation for 'onlineSavingsMsg'
Learn More

CARboxymethyl Cellulose Sodium Salt, Extra Pure, SLR, Low Viscosity, Fisher Chemical

Product Code. 10458590 Shop All Fisher Chemical Products
Change view
Click to view available options
500g, Plastic powder jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10458590 500g, Plastic powder jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10458590 Supplier Fisher Chemical Supplier No. C/3100/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 9004-32-4
Color Yellow
pH 6 to 8
Physical Form Solid
Quantity 500 g
Molecular Formula (C12 H14 O9 R6)n
MDL Number MFCD00081472
InChI Key DPXJVFZANSGRMM-UHFFFAOYNA-N
SMILES [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
Molecular Weight (g/mol) 263.20
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.