missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethyl Cellulose, Sodium Salt, MP Biomedicals™

Product Code. 11477861 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
3 kg
Unit Size:
3kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477861 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11477861 Supplier MP Biomedicals Supplier No. 0215056192

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used as a suspending agent, stabilizer and viscosity-increasing agent. Carboxymethyl Cellulose, Sodium Salt, MP Biomedicals is a high viscosity carboxymethylcellulose (CMC). The viscosity is both concentration and temperature dependent.

The viscosity of is CMC is both concentration and temperature dependent. The viscosity of a 1% solution in water at 25°C is 1300-2200 centipoise (cps). As the temperature increases, the viscosity decreases. As the concentration increases, the viscosity increases.

CMC has a wide variety of industrial, manufacturing and pharmacological uses. It is used as a suspending agent and viscosity modifier (thickener) to stabilize emulsions. Its high viscosity is used to make a mixture which resembles a cream or lotion.

  • In drilling muds
  • In detergents as a soil-suspending agent
  • As a chemical dispersant of oils and other carbon structures such as nanotubes
  • In resin emulsion paints, adhesives, printing inks, textile sizes
  • As protective colloid in general
  • As stabilizer in foods
  • In pharmaceuticals as a suspending agent, tablet excipient and viscosity-increasing agent
  • In the development of biostructures for drug delivery, such as biofilms, emulsions and nanoparticles
TRUSTED_SUSTAINABILITY

Specifications

CAS 9004-32-4
Physical Form Powder
Quantity 3 kg
Molecular Formula (C12 H14 O9 R6)n
MDL Number MFCD00081472
Synonym carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn
Solubility Information Soluble in aqueous solution. High viscosity CMC is soluble at up to 50mg/mL concentration but heat may be required.
InChI Key DPXJVFZANSGRMM-UHFFFAOYNA-N
SMILES [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
Molecular Weight (g/mol) 263.20
PubChem CID 23706213
Formula Weight 700kDa
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.