missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethylcellulose sodium salt

Product Code. 11449256
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449256 500 g 500g
11459256 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449256 Supplier Thermo Scientific Alfa Aesar Supplier No. A18105.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 9004-32-4 | (C12 H14 O9 R6)n | 263.20 g/mol

Carboxymethylcellulose sodium salt is used in drilling muds, in detergents as a soil-suspending agent, in resin emulsion paints, adhesives, printing inks, textile sizes and protective colloid. It acts as a stabilizer in foods. It is also employed in pharmaceuticals as a suspending agent and excipients for tablets. It is used as viscosity modifiers to stabilize the emulsions. It is used as a lubricant in artificial tears and it is used to characterize enzyme activity from endoglucanases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carboxymethylcellulose sodium salt is used in drilling muds, in detergents as a soil-suspending agent, in resin emulsion paints, adhesives, printing inks, textile sizes and protective colloid. It acts as a stabilizer in foods. It is also employed in pharmaceuticals as a suspending agent and excipients for tablets. It is used as viscosity modifiers to stabilize the emulsions. It is used as a lubricant in artificial tears and it is used to characterize enzyme activity from endoglucanases.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-32-4
Molecular Formula (C12 H14 O9 R6)n
Molecular Weight (g/mol) 263.20
MDL Number MFCD00081472
InChI Key DPXJVFZANSGRMM-UHFFFAOYNA-N
Synonym carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn
PubChem CID 23706213
IUPAC Name sodium;2,3,4,5,6-pentahydroxyhexanal;acetate
SMILES [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O

Specifications

Melting Point >300°C
Quantity 500 g
Merck Index 14,1829
Solubility Information Soluble in water.
Chemical Name or Material Carboxymethylcellulose sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.