missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)

Product Code. 10449871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10449871 100 g Glass bottle 100g
10717412 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10449871 Supplier Thermo Scientific Chemicals Supplier No. 332611000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9004-32-4 | (C12 H14 O9 R6)n | 263.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-32-4
Molecular Formula (C12 H14 O9 R6)n
Molecular Weight (g/mol) 263.20
MDL Number MFCD00081472
InChI Key DPXJVFZANSGRMM-UHFFFAOYNA-N
Synonym carboxymethylcellulose sodium usp, celluvisc tn, carmellose sodium jp17, sodium dextrose acetate, c.m.c. tn
PubChem CID 23706213
SMILES [Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O

Specifications

Color White to Beige
pH 6.5 to 8 (1% soln.)
Physical Form Crystals
Quantity 100 g
Infrared Spectrum Authentic
Loss on Drying 10% max. (as packed) (3 to 5 g, 105°C, 2 hrs)
Packaging Glass bottle
Merck Index 15, 1830
Viscosity 2300mPa·s, 2% sol.
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
Percent Purity ≥99.5%
Chemical Name or Material Carboxymethylecellulose, sodium salt, Average M.W. 250000 (DS=0.7)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.