Learn More
ratiolab™ Cardboard Grid Insert for Cryo Boxes
Grid inserts made of ultra low temperature-resistant cardboard.
Brand:  ratiolab™ 5120206
 View more versions of this product
Product Code. 10003063
  • 31.75 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For sample storage, even at ultra-low temperatures
  • Temperature-resistant down to -80°C
  • Water-repellent finish
  • Packaging in practical folding boxes
Specifications
Specifications
10 Inserts/Pk.
Box Divider Insert
7 mm Dia. Tubes
133 mm
Cardboard
40 mm
133 mm
14 x 14