missing translation for 'onlineSavingsMsg'
Learn More

Carfentrazone, TRC

Product Code. 30136892
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136892 50 mg 50mg
30132650 25 mg 25mg
17981505 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136892 Supplier Toronto Research Chemicals Supplier No. TRCC18346550MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128621-72-7
Molecular Formula C13 H10 Cl2 F3 N3 O3
Molecular Weight (g/mol) 384.14
Synonym α,2-Dichloro-5-[4-(difluoromethyl)-4,5-dihydro-3-methyl-5-oxo-1H-1,2,4-triazol-1-yl]-4-fluorobenzenepropanoic acid, 2-Chloro-3-[2-chloro-5-[4-(difluoromethyl)-4,5-dihydro-3-methyl-5-oxo-1H-1,2,4-triazol-1-yl]-4-fluorophenyl]propionic acid, Aim EC, Carfentrazone, Oratio
IUPAC Name 2-chloro-3-[2-chloro-5-[4-(difluoromethyl)-3-methyl-5-oxo-1,2,4-triazol-1-yl]-4-fluorophenyl]propanoic acid
SMILES CC1=NN(C(=O)N1C(F)F)c2cc(CC(Cl)C(=O)O)c(Cl)cc2F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H10Cl2F3N3O3/c1-5-19-21(13(24)20(5)12(17)18)10-3-6(2-8(15)11(22)23)7(14)4-9(10)16/h3-4,8,12H,2H2,1H3,(H,22,23)
Formula Weight 383.0051
Percent Purity >95%
Chemical Name or Material Carfentrazone (free acid)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.