missing translation for 'onlineSavingsMsg'
Learn More

Carminic Acid, >70%, TRC

Product Code. 30352016
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30352016 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30352016

Brand: Toronto Research Chemicals TRCC1836201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carminic Acid
CAS 1260-17-9
Molecular Formula C22 H20 O13
Quantity 1 g
Synonym 2-Anthroic acid, 7-D-glucopyranosyl-9,10-dihydro-3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo- (7CI), Carminic acid (6CI,8CI), 7-β-D-Glucopyranosyl-9,10-dihydro-3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-2-anthracenecarboxylic acid, C.I. 75470, C.I. Natural Red 4, Carmine 5297, Cochineal Red PWD, Creasperse Carmine CP 057, E 120, E 120 (dye), NSC 326224, Natural Red 2180, Natural Red 4, San Red 1, San-Ei Gen San Red 1, Sun Red 1, Sun Red No. 1
InChI Formula InChI=1S/C22H20O13/c1-4-8-5(2-6(24)9(4)22(33)34)13(25)10-11(15(8)27)16(28)12(18(30)17(10)29)21-20(32)19(31)14(26)7(3-23)35-21/h2,7,14,19-21,23-24,26,28-32H,3H2,1H3,(H,33,34)/t7-,14-,19+,20-,21+/m1/s1
SMILES Cc1c(C(=O)O)c(O)cc2C(=O)c3c(O)c(O)c([C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c(O)c3C(=O)c12
IUPAC Name 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid
Molecular Weight (g/mol) 492.39
Formula Weight 492.0904
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.