missing translation for 'onlineSavingsMsg'
Learn More

β-Carotene, MedChemExpress

Product Code. 16484827
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484827 50 mg 50mg
16429076 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16484827 Supplier Medchem Express Supplier No. HYN0411
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress β-Carotene (Provitamin A), a carotenoid compound, is a naturally-occurring vitamin A precursor.

β-Carotene is a modulator of reactive oxygen species (ROS), with antioxidant and antiinflammatory activities. β-Carotene may serve as an antioxidant or as a prooxidant, depending on its intrinsic properties as well as on the redox potential of the biological environment in which it acts. β-Carotene induces breast cancer cells apoptosis, with anticancer activities.

Specifications

Chemical Name or Material β-Carotene
CAS 7235-40-7
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C40H56
Quantity 50 mg
Synonym Provitamin A beta-Carotene
Solubility Information THF : 20 mg/mL (37.25 mM; Need ultrasonic) ∣Acetone : 5 mg/mL (9.31 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.86 mM; ultrasonic and warming and heat to 60°C)
SMILES CC1(C)C(/C=C/C(C)=C/C=C/C(C)=C/C=C/C=C(C)/C=C/C=C(C)/C=C/C2=C(C)CCCC2(C)C)=C(C)CCC1
Molecular Weight (g/mol) 536.87
Formula Weight 536.87
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.