missing translation for 'onlineSavingsMsg'
Learn More

Carprofen, MedChemExpress

Product Code. 16458696
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458696 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458696 Supplier Medchem Express Supplier No. HYB1227
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Carprofen is a nonsteroid anti-inflammatory agent, acts as a multi-target FAAH/COX inhibitor, with IC50s of 3.9 μM, 22.3 μM and 78.6 μM for COX-2, COX-1 and FAAH, respectively.

Carprofen is a nonsteroid anti-inflammatory agent, acts as a multi-target FAAH/COX inhibitor, with IC50s of 3.9 μM, 22.3 μM and 78.6 μM for COX-2, COX-1 and FAAH, respectively.

Specifications

Chemical Name or Material Carprofen
CAS 53716-49-7
Color White
Purity Grade Notes Research
Molecular Formula C15H12ClNO2
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (365.35 mM; Need ultrasonic)
SMILES O=C(O)C(C)C1=CC(NC2=C3C=C(Cl)C=C2)=C3C=C1
Molecular Weight (g/mol) 273.71
Formula Weight 273.71
Percent Purity 99.68%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.