missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Carrying Kit Set for Eutech™ PC 650 meter
Description
Eutech™ Carrying Kit Cases are thoughtfully designed to hold Eutech™ meters and electrodes.The Carrying Kit Set is a plastic carrying case for the Eutech™ PC 650 meter and probe, calibration solutions, batteries, and manuals.
The Carrying Kit set is comprised of:
- pH 4.0 Buffer Solution
Specifications
Specifications
| Description | Carrying Kit Set for Eutech™ PC 650 meter |
| Type | Accessory |
| Includes | pH 4.0 Buffer Solution, pH 7.00 Buffer Solution, 12.88 mS/cm KCl Calibration Solution, Deionised rinse water |
| For Use With (Equipment) | Eutech™ PC 650 meter |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?