missing translation for 'onlineSavingsMsg'
Learn More

Usbeck Laborgeraete™ Cartridge Burner with safety element

Product Code. 10219201
Change view
Click to view available options
Gas Type:
Butane, LPG, Propane, Propane
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gas Type unitSize
10219201 Butane, LPG, Propane, Propane Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10219201 Supplier Usbeck Laborgeraete™ Supplier No. 1425
  • ONLINE EXCLUSIVE PRICE
    108.00 €
    106.55 € / Pack of 1
    Save 1.45 €
    1% Off
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Usbeck Laborgeraete Cartridge Burner with Safety Element - Provides protection of uncontrolled escape of gas in laboratories.

  • Din Type
  • Needle Valve
  • Includes 400 mL Cartridge
  • T = 1650°C
  • Suitable Tripod: 5136/5146
  • Provides protection of uncontrolled escape of gas in laboratories.
  • By the heat of the pilot flame, an electric voltage is generated within the thermocouple. Due to this voltage, a magnetic valve in the safety element keeps open. The gas is able to flow.
  • In case the flame of te burners extinguishes, the thermocouple cools down and the electric voltage drops off, thus closing the magnetic valve automatically. The gas supply is interrupted and no unburnt gas can escape.

Specifications

Product Type Gas Cartridge
Gas Type Butane, LPG, Propane, Propane
Includes 400 mL Cartridge
Max. Temperature (Metric) 1650°C
Safety Features Magnetic Valve in the Safety Element
compliance-icons
Product Identifier
  • Cartridge burnerwithsafety element, DIN type .
Signal Word
  • Danger
Hazard Category
  • Flammable gas.
  • Gases under pressure
Hazard Statement
  • H220-Extremely flammable gas.
  • H280-Contains gas under pressure; may explode if heated.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P377-Leaking gas fire: Do not extinguish, unless leak can be stopped safely.
  • P381-Eliminate all ignition sources if safe to do so.
  • P403-Store in a well-ventilated place.
Supplemental information
  • MIXTURE LIST-Contain: n-Butane, Propane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.