missing translation for 'onlineSavingsMsg'
Learn More

β-Caryophyllene, MedChemExpress

Product Code. 16432396
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16432396 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16432396 Supplier Medchem Express Supplier No. HYN1415
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress β-Caryophyllene is a CB2 receptor agonist.

β-Caryophyllene is a CB2 receptor agonist.

Specifications

Chemical Name or Material β-Caryophyllene
CAS 87-44-5
Color Colorless
Purity Grade Notes Research
Molecular Formula C15H24
Quantity 500 mg
Synonym (-)-(E)-Caryophyllene (−)-β-caryophyllene (−)-trans-Caryophyllene
Solubility Information Ethanol : ≥ 176.67 mg/mL (864.55 mM) ∣DMSO : 50 mg/mL (244.68 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 0.67 mg/mL (3.28 mM; Need ultrasonic)
SMILES C=C1CC/C=C(C)/CC[C@@]2([H])C(C)(C)C[C@]12[H]
Molecular Weight (g/mol) 204.35
Formula Weight 204.35
Percent Purity 95.0%
Grade Research
Physical Form Liquid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.